This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC00844 SIRNA Assays ›
Gene Symbol:
LINC00844
Gene Name:
long intergenic non-protein coding RNA 844
Gene Aliases:
-
Chromosome Location:
Chr.10: 58999518 - 59001617 on Build GRCh38
UniGene ID:
Hs.170953
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n497671

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC00844
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_108046.1 2 396
GenBank mRNA CB157564.1 - 2 280
DA153602.1 - 2 313
DB500612.1 - 2 397
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA469334.1 - 2 280 Chr. 10 - 58999639 - 59001435 [+] on Build GRCh38
GenBankmRNA AV694540.1 - 274 Chr. - - [-] on Build
GenBankmRNA DA386826.1 - 2 316 Chr. 10 - 58999598 - 59001616 [+] on Build GRCh38
GenBankmRNA DB571782.1 - 2 362 Chr. 10 - 58999552 - 59001470 [+] on Build GRCh38
GenBankmRNA BG618474.1 - 290 Chr. - - [-] on Build
GenBankmRNA BX096738.1 - 2 282 Chr. 10 - 58999632 - 59001471 [+] on Build GRCh38
GenBankmRNA BF680613.1 - 284 Chr. - - [-] on Build
GenBankmRNA BM714416.1 - 2 251 Chr. 10 - 58999663 - 59001471 [+] on Build GRCh38
GenBankmRNA DA116548.1 - 2 355 Chr. 10 - 58999559 - 59001559 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0