siRNA Details
Gene Symbol: LOC101928098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132786.1 | 2 | 687 | |||
GenBank mRNA | BC040907.1 | - | 2 | 687 |

Gene Symbol: LOC101928098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132786.1 | 2 | 687 | |||
GenBank mRNA | BC040907.1 | - | 2 | 687 |