siRNA Details
Gene Symbol: LOC101928307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125872.1 | 2 | 495 | |||
GenBank mRNA | AB073651.1 | - | 1 | 2165 | ||
AL833240.1 | - | 1 | 2328 | |||
BM686538.1 | - | 2 | 503 |

Gene Symbol: LOC101928307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125872.1 | 2 | 495 | |||
GenBank mRNA | AB073651.1 | - | 1 | 2165 | ||
AL833240.1 | - | 1 | 2328 | |||
BM686538.1 | - | 2 | 503 |