siRNA Details
Gene Symbol: VIM-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108060.1 | 4 | 1023 | |||
NR_108061.1 | 3 | 1547 |

Gene Symbol: VIM-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108060.1 | 4 | 1023 | |||
NR_108061.1 | 3 | 1547 |