siRNA Details
Gene Symbol: 2400006E01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019103.1 | - | 2 | 285 | ||
AV500291.1 | - | 1 | 182 |

Gene Symbol: 2400006E01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019103.1 | - | 2 | 285 | ||
AV500291.1 | - | 1 | 182 |