This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALS2CR11 SIRNA Assays ›
Gene Symbol:
Als2cr11
Gene Name:
amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11 (human)
Gene Aliases:
1700052H20Rik, 4930408G06Rik
Chromosome Location:
Chr.1: 59050506 - 59095015 on Build GRCm38
UniGene ID:
Mm.252811
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n441441

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Als2cr11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK015211.1 - 364
AK019575.1 - 474
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CN834703.1 - 247 Chr. - - [-] on Build
GenBankmRNA BY714657.1 - 2 474 Chr. 1 - 58997326 - 59003539 [-] on Build GRCm38
GenBankmRNA BY714766.1 - 2 364 Chr. 1 - 58997117 - 59003429 [-] on Build GRCm38
GenBankmRNA CN837599.1 - 2 213 Chr. 1 - 58997142 - 59003278 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0