siRNA Details
Gene Symbol: A530065N20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046142.1 | 3 | 1083 | |||
GenBank mRNA | AK080130.1 | - | 3 | 1086 |

Gene Symbol: A530065N20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046142.1 | 3 | 1083 | |||
GenBank mRNA | AK080130.1 | - | 3 | 1086 |