This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other A030001D20RIK SIRNA Assays ›
Gene Symbol:
A030001D20Rik
Gene Name:
RIKEN cDNA A030001D20 gene
Gene Aliases:
1110025D03Rik
Chromosome Location:
-
UniGene ID:
Mm.440600
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n440689

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: A030001D20Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK003910.1 - 2 275
AK037147.1 - 2 291
AK049852.1 - 2 277
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB630270.1 - 2 291 Chr. 7 - 45636556 - 45638758 [-] on Build GRCm38
GenBankmRNA CK031458.1 - 2 275 Chr. 7 - 45636472 - 45638736 [-] on Build GRCm38
GenBankmRNA BB655923.1 - 2 288 Chr. 7 - 45636558 - 45638755 [-] on Build GRCm38
GenBankmRNA CX202519.1 - 2 282 Chr. 7 - 45636407 - 45638750 [-] on Build GRCm38
GenBankmRNA AA734726.1 - 2 277 Chr. 7 - 45636793 - 45638744 [-] on Build GRCm38
GenBankmRNA CB589422.1 - 2 297 Chr. 7 - 45636424 - 45638765 [-] on Build GRCm38
GenBankmRNA BU561856.1 - 284 Chr. - - [-] on Build
GenBankmRNA BY704165.1 - 2 275 Chr. 7 - 45636217 - 45638742 [-] on Build GRCm38
GenBankmRNA BB597893.2 - 2 277 Chr. 7 - 45636547 - 45638744 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0