This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM118B SIRNA Assays ›
Gene Symbol:
Fam118b
Gene Name:
family with sequence similarity 118, member B
Gene Aliases:
2310022O21Rik, 2700018L24Rik, C030004A17Rik, Ppp4r1l
Chromosome Location:
Chr.9: 35216965 - 35267812 on Build GRCm38
UniGene ID:
Mm.24197
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n439893

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam118b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK054309.1 - 1455
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB554517.2 - 1 214 Chr. 1 - 65017970 - 65018755 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0