This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM24105 SIRNA Assays ›
Gene Symbol:
Gm24105
Gene Name:
predicted gene, 24105
Gene Aliases:
-
Chromosome Location:
Chr.5: 149236812 - 149248644 on Build GRCm38
UniGene ID:
Mm.366544
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n436703

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm24105
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK155259.1 - 88
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY748885.1 - 1 84 Chr. 5 - 149236731 - 149247679 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-6889b944d9-flx6z:80/100.66.128.142:80.
git-commit: df865406a845caddbd23a531a4b79e504db49c75
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header