This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM16096 SIRNA Assays ›
Gene Symbol:
Gm16096
Gene Name:
predicted gene 16096
Gene Aliases:
OTTMUSG00000030243
Chromosome Location:
Chr.9: 40782611 - 40786468 on Build GRCm38
UniGene ID:
Mm.444560
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n434045

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm16096
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK089373.1 - 251
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY749634.1 - 2 251 Chr. 9 - 40780879 - 40784181 [-] on Build GRCm38
GenBankmRNA AV454648.1 - 2 204 Chr. 9 - 40780990 - 40784129 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0