This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1810021B22RIK SIRNA Assays ›
Gene Symbol:
1810021B22Rik
Gene Name:
RIKEN cDNA 1810021B22 gene
Gene Aliases:
-
Chromosome Location:
Chr.15: 89071198 - 89075854 on Build GRCm38
UniGene ID:
Mm.276391
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n431951

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1810021B22Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040417.1 3 442
GenBank mRNA AK007571.1 - 3 443
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY707618.1 - 3 437 Chr. 15 - 89071196 - 89075852 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0