This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM12718 SIRNA Assays ›
Gene Symbol:
Gm12718
Gene Name:
predicted gene 12718
Gene Aliases:
OTTMUSG00000008109
Chromosome Location:
Chr.4: 103382500 - 103492188 on Build GRCm38
UniGene ID:
Mm.244115
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n431661

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm12718
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040673.1 1 16
GenBank mRNA AK144477.1 - 1 17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY733749.1 - 1 17 Chr. 4 - 103382500 - 103492040 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header