siRNA Details
Gene Symbol: 4930548J01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045462.1 | 2 | 171 | |||
GenBank mRNA | AK007199.1 | - | 2 | 183 | ||
AK019751.1 | - | 2 | 172 |

Gene Symbol: 4930548J01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045462.1 | 2 | 171 | |||
GenBank mRNA | AK007199.1 | - | 2 | 183 | ||
AK019751.1 | - | 2 | 172 |