siRNA Details
Gene Symbol: 4930421P05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015172.1 | - | 2 | 397 | ||
AK077193.1 | - | 1 | 1241 |

Gene Symbol: 4930421P05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015172.1 | - | 2 | 397 | ||
AK077193.1 | - | 1 | 1241 |