siRNA Details
Gene Symbol: 4933417C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA536659.1 | - | 1 | 455 | ||
AK076659.1 | - | 1 | 463 | |||
AV259421.2 | - | 1 | 180 |

Gene Symbol: 4933417C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA536659.1 | - | 1 | 455 | ||
AK076659.1 | - | 1 | 463 | |||
AV259421.2 | - | 1 | 180 |