This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FNBP1 SIRNA Assays ›
Gene Symbol:
Fnbp1
Gene Name:
formin binding protein 1
Gene Aliases:
1110057E06Rik, 2210010H06Rik, FBP1, Fbp17
Chromosome Location:
Chr.2: 31026206 - 31142027 on Build GRCm38
UniGene ID:
Mm.440993
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n417678

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fnbp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK041983.1 - 1 1693
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CK625939.1 - 1 123 Chr. 2 - 31029128 - 31029690 [-] on Build GRCm38
GenBankmRNA AA184890.1 - 1 43 Chr. 2 - 31029334 - 31029598 [-] on Build GRCm38
GenBankmRNA CB318832.1 - 1 56 Chr. 2 - 31029265 - 31029621 [-] on Build GRCm38
GenBankmRNA BB234652.2 - 1 408 Chr. 2 - 31029286 - 31029976 [-] on Build GRCm38
GenBankmRNA BX525913.1 - 1 38 Chr. 2 - 31029267 - 31029598 [-] on Build GRCm38
GenBankmRNA CK626428.1 - 1 534 Chr. 2 - 31029479 - 31030101 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0