This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PBX1 SIRNA Assays ›
Gene Symbol:
Pbx1
Gene Name:
pre B cell leukemia homeobox 1
Gene Aliases:
2310056B04Rik, D230003C07Rik, Pbx-1
Chromosome Location:
Chr.1: 168119364 - 168432264 on Build GRCm38
UniGene ID:
Mm.43358
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n414644

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pbx1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK083616.1 - 1 1458
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY467573.1 - 1 171 Chr. 1 - 168153551 - 168153929 [-] on Build GRCm38
GenBankmRNA CJ258330.1 - 1 232 Chr. 1 - 168153551 - 168153989 [-] on Build GRCm38
GenBankmRNA BX517377.1 - 1 434 Chr. 1 - 168153678 - 168154195 [-] on Build GRCm38
GenBankmRNA AV015754.1 - 1 131 Chr. 1 - 168153551 - 168153886 [-] on Build GRCm38
GenBankmRNA BG087478.2 - 1 266 Chr. 1 - 168153552 - 168154023 [-] on Build GRCm38
GenBankmRNA BB537829.1 - 1 74 Chr. 1 - 168153551 - 168153831 [-] on Build GRCm38
GenBankmRNA BB553390.1 - 1 65 Chr. 1 - 168153551 - 168153789 [-] on Build GRCm38
GenBankmRNA BB446070.2 - 1 416 Chr. 1 - 168153527 - 168154173 [-] on Build GRCm38
GenBankmRNA CB248204.1 - 1 660 Chr. 1 - 168153590 - 168154417 [-] on Build GRCm38
GenBankmRNA BE981671.1 - 1 170 Chr. 1 - 168153547 - 168153927 [-] on Build GRCm38
GenBankmRNA CJ258792.1 - 1 298 Chr. 1 - 168153551 - 168154055 [-] on Build GRCm38
GenBankmRNA DV552784.1 - 1 225 Chr. 1 - 168153529 - 168153981 [-] on Build GRCm38
GenBankmRNA BE634095.1 - 1 184 Chr. 1 - 168153548 - 168153941 [-] on Build GRCm38
GenBankmRNA BB392910.1 - 1 121 Chr. 1 - 168153551 - 168153878 [-] on Build GRCm38
GenBankmRNA BB532528.1 - 1 127 Chr. 1 - 168153551 - 168153884 [-] on Build GRCm38
GenBankmRNA BB228846.1 - 1 97 Chr. 1 - 168153551 - 168153855 [-] on Build GRCm38
GenBankmRNA DV047611.1 - 243 Chr. - - [-] on Build
GenBankmRNA BF466686.1 - 1 120 Chr. 1 - 168153580 - 168153877 [-] on Build GRCm38
GenBankmRNA CJ259288.1 - 1 245 Chr. 1 - 168153551 - 168154002 [-] on Build GRCm38
GenBankmRNA BB027467.1 - 1 78 Chr. 1 - 168153551 - 168153835 [-] on Build GRCm38
GenBankmRNA CN533958.1 - 1 166 Chr. 1 - 168153532 - 168153884 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0