siRNA Details
Gene Symbol: 3110015C05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045908.1 | 3 | 880 | |||
GenBank mRNA | AK014052.1 | - | 3 | 880 |

Gene Symbol: 3110015C05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045908.1 | 3 | 880 | |||
GenBank mRNA | AK014052.1 | - | 3 | 880 |