This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4930402F11RIK SIRNA Assays ›
Gene Symbol:
4930402F11Rik
Gene Name:
RIKEN cDNA 4930402F11 gene
Gene Aliases:
-
Chromosome Location:
Chr.7: 69496094 - 69499689 on Build GRCm38
UniGene ID:
Mm.410832
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n412037

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930402F11Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045940.1 2, 3 145
GenBank mRNA AK015048.1 - 2, 3 147
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY714574.1 - 147 Chr. - - [-] on Build

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0