siRNA Details
Gene Symbol: 4930539M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040597.1 | 3 | 653 | |||
GenBank mRNA | AK016002.1 | - | 3 | 660 | ||
BB615931.1 | - | 3 | 653 |

Gene Symbol: 4930539M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040597.1 | 3 | 653 | |||
GenBank mRNA | AK016002.1 | - | 3 | 660 | ||
BB615931.1 | - | 3 | 653 |