This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM4710 SIRNA Assays ›
Gene Symbol:
Gm4710
Gene Name:
predicted gene 4710
Gene Aliases:
-
Chromosome Location:
Chr.17: 75861023 - 75889039 on Build GRCm38
UniGene ID:
Mm.127562
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n408426

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm4710
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033456.1 3 465
GenBank mRNA AK082440.1 - 3 466
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB650123.1 - 3 466 Chr. 17 - 75861023 - 75886387 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header