siRNA Details
Gene Symbol: 4930556M19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045063.1 | 7 | 1367 | |||
GenBank mRNA | AK030258.1 | - | 7 | 1368 |

Gene Symbol: 4930556M19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045063.1 | 7 | 1367 | |||
GenBank mRNA | AK030258.1 | - | 7 | 1368 |