siRNA Details
Gene Symbol: 4922502N22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045149.1 | 4 | 591 | |||
GenBank mRNA | AK015023.1 | - | 4 | 593 | ||
AK132719.1 | - | 4 | 593 |

Gene Symbol: 4922502N22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045149.1 | 4 | 591 | |||
GenBank mRNA | AK015023.1 | - | 4 | 593 | ||
AK132719.1 | - | 4 | 593 |