siRNA Details
Gene Symbol: A530006G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046014.1 | 3 | 1316 | |||
GenBank mRNA | AK079921.1 | - | 3 | 1317 |

Gene Symbol: A530006G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046014.1 | 3 | 1316 | |||
GenBank mRNA | AK079921.1 | - | 3 | 1317 |