This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other A530006G24RIK SIRNA Assays ›
Gene Symbol:
A530006G24Rik
Gene Name:
RIKEN cDNA A530006G24 gene
Gene Aliases:
Gm1003
Chromosome Location:
Chr.2: 147710693 - 147717632 on Build GRCm38
UniGene ID:
Mm.259809
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n404312

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: A530006G24Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_046014.1 3 338
GenBank mRNA AK079921.1 - 3 339
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB636516.1 - 3 339 Chr. 2 - 147710693 - 147716947 [+] on Build GRCm38
GenBankmRNA BB636075.1 - 3 339 Chr. 2 - 147710693 - 147716925 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0