This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LARP4B SIRNA Assays ›
Gene Symbol:
Larp4b
Gene Name:
La ribonucleoprotein domain family, member 4B
Gene Aliases:
A630096F19, AI256361, D13Wsu64e, Larp5
Chromosome Location:
Chr.13: 9093814 - 9174406 on Build GRCm38
UniGene ID:
Mm.234944
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n404178

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Larp4b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK134899.1 - 1 1181
AK172916.1 - 19 4921
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA EH097810.1 - 2 291 Chr. 13 - 9172180 - 9174309 [+] on Build GRCm38
GenBankmRNA EH100785.1 - 1 378 Chr. 13 - 9173825 - 9174406 [+] on Build GRCm38
GenBankmRNA AA407164.1 - 1 189 Chr. 13 - 9174005 - 9174364 [+] on Build GRCm38
GenBankmRNA AA467521.1 - 1 294 Chr. 13 - 9173898 - 9174402 [+] on Build GRCm38
GenBankmRNA AA409805.1 - 1 189 Chr. 13 - 9174005 - 9174406 [+] on Build GRCm38
GenBankmRNA CK031178.1 - 1 277 Chr. 13 - 9173931 - 9174404 [+] on Build GRCm38
GenBankmRNA BB825695.1 - 1 249 Chr. 13 - 9173951 - 9174404 [+] on Build GRCm38
GenBankmRNA AA823532.1 - 1 148 Chr. 13 - 9174049 - 9174375 [+] on Build GRCm38
GenBankmRNA AA254632.1 - 1 297 Chr. 13 - 9173897 - 9174406 [+] on Build GRCm38
GenBankmRNA BY405476.1 - 1 203 Chr. 13 - 9173991 - 9174404 [+] on Build GRCm38
GenBankmRNA BE135693.1 - 1 258 Chr. 13 - 9173951 - 9174284 [+] on Build GRCm38
GenBankmRNA AA797681.1 - 1 369 Chr. 13 - 9173826 - 9174258 [+] on Build GRCm38
GenBankmRNA BY604810.1 - 1 179 Chr. 13 - 9174015 - 9174445 [+] on Build GRCm38
GenBankmRNA CK128165.1 - 1 392 Chr. 13 - 9173802 - 9174403 [+] on Build GRCm38
GenBankmRNA BE635007.1 - 1 355 Chr. 13 - 9173839 - 9174303 [+] on Build GRCm38
GenBankmRNA AA212701.1 - 1 255 Chr. 13 - 9173939 - 9174370 [+] on Build GRCm38
GenBankmRNA EH111403.1 - 1 94 Chr. 13 - 9174100 - 9174402 [+] on Build GRCm38
GenBankmRNA DV047109.1 - 183 Chr. - - [-] on Build

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0