This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1700020I14RIK SIRNA Assays ›
Gene Symbol:
1700020I14Rik
Gene Name:
RIKEN cDNA 1700020I14 gene
Gene Aliases:
1190006L01Rik, 4933437I03Rik, AI451541, AI585852, AW538376, AW558897, Cyrano
Chromosome Location:
Chr.2: 119594296 - 119600746 on Build GRCm38
UniGene ID:
Mm.379181
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n403538

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700020I14Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK004510.1 - 1108
AK147270.1 - 7943
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AV459727.1 - 1 268 Chr. 2 - 119606891 - 119607339 [+] on Build GRCm38
GenBankmRNA CJ258093.1 - 1 145 Chr. 2 - 119607014 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ259920.1 - 1 91 Chr. 2 - 119607068 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ251706.1 - 1 153 Chr. 2 - 119607006 - 119607494 [+] on Build GRCm38
GenBankmRNA BF147459.1 - 1 109 Chr. 2 - 119607050 - 119607378 [+] on Build GRCm38
GenBankmRNA CJ252811.1 - 1 126 Chr. 2 - 119607033 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ258930.1 - 1 113 Chr. 2 - 119607046 - 119607494 [+] on Build GRCm38
GenBankmRNA BX514013.1 - 2 320 Chr. 2 - 119256504 - 119607341 [+] on Build GRCm38
GenBankmRNA CJ261249.1 - 1 117 Chr. 2 - 119607042 - 119607497 [+] on Build GRCm38
GenBankmRNA CJ258411.1 - 1 111 Chr. 2 - 119607048 - 119607494 [+] on Build GRCm38
GenBankmRNA BE626961.1 - 1 178 Chr. 2 - 119606981 - 119607497 [+] on Build GRCm38
GenBankmRNA BB751543.1 - 1 180 Chr. 2 - 119606979 - 119607490 [+] on Build GRCm38
GenBankmRNA CJ258729.1 - 1 164 Chr. 2 - 119606995 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ256132.1 - 1 137 Chr. 2 - 119607022 - 119607494 [+] on Build GRCm38
GenBankmRNA AA178638.1 - 1 286 Chr. 2 - 119606873 - 119607340 [+] on Build GRCm38
GenBankmRNA CJ254189.1 - 1 135 Chr. 2 - 119607024 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ253559.1 - 1 106 Chr. 2 - 119607053 - 119607492 [+] on Build GRCm38
GenBankmRNA CJ261355.1 - 1 137 Chr. 2 - 119607022 - 119607494 [+] on Build GRCm38
GenBankmRNA DT918331.1 - 1 335 Chr. 2 - 119606824 - 119607487 [+] on Build GRCm38
GenBankmRNA CJ257461.1 - 1 212 Chr. 2 - 119606948 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ255911.1 - 1 107 Chr. 2 - 119607052 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ253031.1 - 1 166 Chr. 2 - 119606993 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ262718.1 - 1 154 Chr. 2 - 119607006 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ255899.1 - 1 118 Chr. 2 - 119607041 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ256917.1 - 1 167 Chr. 2 - 119606992 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ250421.1 - 1 145 Chr. 2 - 119607014 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ256212.1 - 1 140 Chr. 2 - 119607020 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ254937.1 - 1 135 Chr. 2 - 119607025 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ261973.1 - 1 141 Chr. 2 - 119607018 - 119607494 [+] on Build GRCm38
GenBankmRNA CK128973.1 - 1 349 Chr. 2 - 119606810 - 119607497 [+] on Build GRCm38
GenBankmRNA BG088876.2 - 1 234 Chr. 2 - 119606925 - 119607493 [+] on Build GRCm38
GenBankmRNA CB196004.1 - 372 Chr. - - [-] on Build
GenBankmRNA CJ244259.1 - 1 74 Chr. 2 - 119607085 - 119607497 [+] on Build GRCm38
GenBankmRNA CJ249575.1 - 1 155 Chr. 2 - 119607004 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ260937.1 - 1 106 Chr. 2 - 119607053 - 119607494 [+] on Build GRCm38
GenBankmRNA AI121742.1 - 1 68 Chr. 2 - 119607091 - 119607490 [+] on Build GRCm38
GenBankmRNA CJ244173.1 - 1 62 Chr. 2 - 119607098 - 119607497 [+] on Build GRCm38
GenBankmRNA CJ253043.1 - 1 165 Chr. 2 - 119606994 - 119607494 [+] on Build GRCm38
GenBankmRNA BF661480.1 - 1 105 Chr. 2 - 119607054 - 119607492 [+] on Build GRCm38
GenBankmRNA AI117931.1 - 1 77 Chr. 2 - 119607082 - 119607466 [+] on Build GRCm38
GenBankmRNA CB847061.2 - 1 31 Chr. 2 - 119607139 - 119607497 [+] on Build GRCm38
GenBankmRNA CJ261368.1 - 1 116 Chr. 2 - 119607043 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ262427.1 - 1 191 Chr. 2 - 119606975 - 119607494 [+] on Build GRCm38
GenBankmRNA CJ261784.1 - 1 156 Chr. 2 - 119607003 - 119607494 [+] on Build GRCm38
GenBankmRNA CD804325.1 - 1 101 Chr. 2 - 119607058 - 119607497 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0