This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DPP8 SIRNA Assays ›
Gene Symbol:
Dpp8
Gene Name:
dipeptidylpeptidase 8
Gene Aliases:
2310004I03Rik, 4932434F09Rik, AI666706, DPP VIII
Chromosome Location:
Chr.9: 65032458 - 65084503 on Build GRCm38
UniGene ID:
Mm.202749
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n397685

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dpp8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK009140.1 - 1 983
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB193812.1 - 1 54 Chr. 9 - 65084210 - 65084503 [+] on Build GRCm38
GenBankmRNA AA690500.1 - 1 311 Chr. 9 - 65083950 - 65084501 [+] on Build GRCm38
GenBankmRNA CD350652.1 - 1 386 Chr. 9 - 65083876 - 65084501 [+] on Build GRCm38
GenBankmRNA CX206847.1 - 1 305 Chr. 9 - 65083994 - 65084501 [+] on Build GRCm38
GenBankmRNA BY474602.1 - 1 189 Chr. 9 - 65084074 - 65084501 [+] on Build GRCm38
GenBankmRNA AA210277.1 - 1 251 Chr. 9 - 65084011 - 65084499 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0