This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM13944 SIRNA Assays ›
Gene Symbol:
Gm13944
Gene Name:
predicted gene 13944
Gene Aliases:
OTTMUSG00000015155
Chromosome Location:
Chr.2: 77462644 - 77480938 on Build GRCm38
UniGene ID:
Mm.443215
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n397377

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm13944
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040368.1 2 220
GenBank mRNA AK078367.1 - 2 171
CJ104421.1 - 2 221
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY719255.1 - 2 171 Chr. 2 - 77462694 - 77480868 [+] on Build GRCm38
GenBankmRNA CJ113664.1 - 2 221 Chr. 2 - 77462644 - 77480661 [+] on Build GRCm38
GenBankmRNA CJ112582.1 - 2 221 Chr. 2 - 77462644 - 77480724 [+] on Build GRCm38
GenBankmRNA CJ105133.1 - 2 221 Chr. 2 - 77462644 - 77480627 [+] on Build GRCm38
GenBankmRNA CJ113273.1 - 2 222 Chr. 2 - 77462644 - 77480709 [+] on Build GRCm38
GenBankmRNA CJ116062.1 - 2 221 Chr. 2 - 77462644 - 77480697 [+] on Build GRCm38
GenBankmRNA CJ112676.1 - 2 221 Chr. 2 - 77462644 - 77480682 [+] on Build GRCm38
GenBankmRNA CJ119172.1 - 2 222 Chr. 2 - 77462644 - 77480683 [+] on Build GRCm38
GenBankmRNA CJ112127.1 - 2 221 Chr. 2 - 77462644 - 77480726 [+] on Build GRCm38
GenBankmRNA CJ120144.1 - 2 221 Chr. 2 - 77462644 - 77480707 [+] on Build GRCm38
GenBankmRNA CJ121682.1 - 2 221 Chr. 2 - 77462644 - 77480684 [+] on Build GRCm38
GenBankmRNA CJ115353.1 - 2 221 Chr. 2 - 77462644 - 77480697 [+] on Build GRCm38
GenBankmRNA CJ105865.1 - 2 221 Chr. 2 - 77462644 - 77480679 [+] on Build GRCm38
GenBankmRNA CJ118296.1 - 2 221 Chr. 2 - 77462644 - 77480669 [+] on Build GRCm38
GenBankmRNA CJ107429.1 - 2 221 Chr. 2 - 77462644 - 77480695 [+] on Build GRCm38
GenBankmRNA CJ121271.1 - 2 221 Chr. 2 - 77462644 - 77480664 [+] on Build GRCm38
GenBankmRNA CJ105541.1 - 2 221 Chr. 2 - 77462644 - 77480695 [+] on Build GRCm38
GenBankmRNA CJ122176.1 - 2 221 Chr. 2 - 77462644 - 77480688 [+] on Build GRCm38
GenBankmRNA CJ104590.1 - 2 221 Chr. 2 - 77462644 - 77480672 [+] on Build GRCm38
GenBankmRNA BM936406.1 - 3 353 Chr. 2 - 77462669 - 77480593 [+] on Build GRCm38
GenBankmRNA CJ119674.1 - 2 221 Chr. 2 - 77462644 - 77480672 [+] on Build GRCm38
GenBankmRNA CJ113422.1 - 2 221 Chr. 2 - 77462644 - 77480704 [+] on Build GRCm38
GenBankmRNA CJ106823.1 - 2 221 Chr. 2 - 77462644 - 77480727 [+] on Build GRCm38
GenBankmRNA BY705307.1 - 2 187 Chr. 2 - 77462677 - 77480801 [+] on Build GRCm38
GenBankmRNA CJ109011.1 - 2 221 Chr. 2 - 77462644 - 77480703 [+] on Build GRCm38
GenBankmRNA CJ113744.1 - 2 221 Chr. 2 - 77462644 - 77480679 [+] on Build GRCm38
GenBankmRNA CJ108331.1 - 2 221 Chr. 2 - 77462644 - 77480559 [+] on Build GRCm38
GenBankmRNA CJ117453.1 - 2 221 Chr. 2 - 77462644 - 77480657 [+] on Build GRCm38
GenBankmRNA CJ122742.1 - 2 221 Chr. 2 - 77462644 - 77480669 [+] on Build GRCm38
GenBankmRNA CJ116465.1 - 2 222 Chr. 2 - 77462644 - 77480670 [+] on Build GRCm38
GenBankmRNA CJ104739.1 - 2 221 Chr. 2 - 77462644 - 77480681 [+] on Build GRCm38
GenBankmRNA CJ120892.1 - 2 221 Chr. 2 - 77462644 - 77480678 [+] on Build GRCm38
GenBankmRNA CJ121553.1 - 2 221 Chr. 2 - 77462644 - 77480665 [+] on Build GRCm38
GenBankmRNA CJ108907.1 - 2 221 Chr. 2 - 77462644 - 77480698 [+] on Build GRCm38
GenBankmRNA CJ118328.1 - 2 221 Chr. 2 - 77462644 - 77480466 [+] on Build GRCm38
GenBankmRNA CJ120006.1 - 2 221 Chr. 2 - 77462644 - 77480691 [+] on Build GRCm38
GenBankmRNA CJ114939.1 - 2 221 Chr. 2 - 77462644 - 77480681 [+] on Build GRCm38
GenBankmRNA CJ119477.1 - 2 221 Chr. 2 - 77462644 - 77480686 [+] on Build GRCm38
GenBankmRNA CJ122553.1 - 2 221 Chr. 2 - 77462644 - 77480663 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0