This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM13944 SIRNA Assays ›
Gene Symbol:
Gm13944
Gene Name:
predicted gene 13944
Gene Aliases:
OTTMUSG00000015155
Chromosome Location:
Chr.2: 77462644 - 77480938 on Build GRCm38
UniGene ID:
Mm.443215
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n397376

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm13944
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040368.1 3 491
GenBank mRNA AK078367.1 - 3 442
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CJ113422.1 - 3 495 Chr. 2 - 77462644 - 77480704 [+] on Build GRCm38
GenBankmRNA BY719255.1 - 3 442 Chr. 2 - 77462694 - 77480868 [+] on Build GRCm38
GenBankmRNA BY705307.1 - 3 461 Chr. 2 - 77462677 - 77480801 [+] on Build GRCm38
GenBankmRNA CJ113273.1 - 3 497 Chr. 2 - 77462644 - 77480709 [+] on Build GRCm38
GenBankmRNA CJ109011.1 - 3 495 Chr. 2 - 77462644 - 77480703 [+] on Build GRCm38
GenBankmRNA CJ120144.1 - 3 495 Chr. 2 - 77462644 - 77480707 [+] on Build GRCm38
GenBankmRNA CJ115353.1 - 3 495 Chr. 2 - 77462644 - 77480697 [+] on Build GRCm38
GenBankmRNA CJ107429.1 - 3 495 Chr. 2 - 77462644 - 77480695 [+] on Build GRCm38
GenBankmRNA CJ105541.1 - 3 495 Chr. 2 - 77462644 - 77480695 [+] on Build GRCm38
GenBankmRNA CJ108907.1 - 3 495 Chr. 2 - 77462644 - 77480698 [+] on Build GRCm38
GenBankmRNA CJ120006.1 - 3 495 Chr. 2 - 77462644 - 77480691 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0