This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE2D3 SIRNA Assays ›
Gene Symbol:
Ube2d3
Gene Name:
ubiquitin-conjugating enzyme E2D 3
Gene Aliases:
1100001F19Rik, 9430029A22Rik, AA414951
Chromosome Location:
Chr.3: 135438244 - 135467180 on Build GRCm38
UniGene ID:
Mm.391601
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n396163

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ube2d3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK020443.1 - 260
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AI173870.1 - 1 402 Chr. 3 - 135467460 - 135468063 [+] on Build GRCm38
GenBankmRNA AI119345.1 - 1 411 Chr. 3 - 135467451 - 135467976 [+] on Build GRCm38
GenBankmRNA BB706043.1 - 1 132 Chr. 3 - 135467729 - 135468193 [+] on Build GRCm38
GenBankmRNA BB745642.1 - 1 132 Chr. 3 - 135467730 - 135468193 [+] on Build GRCm38
GenBankmRNA AA710466.1 - 1 77 Chr. 3 - 135467785 - 135468183 [+] on Build GRCm38
GenBankmRNA CF182374.1 - 1 590 Chr. 3 - 135467272 - 135467894 [+] on Build GRCm38
GenBankmRNA AV578136.1 - 1 384 Chr. 3 - 135467477 - 135467930 [+] on Build GRCm38
GenBankmRNA BY721675.1 - 1 256 Chr. 3 - 135467606 - 135468197 [+] on Build GRCm38
GenBankmRNA AA285723.1 - 1 129 Chr. 3 - 135467733 - 135468186 [+] on Build GRCm38
GenBankmRNA BI248421.1 - 1 352 Chr. 3 - 135467510 - 135468198 [+] on Build GRCm38
GenBankmRNA CJ312772.1 - 1 187 Chr. 3 - 135467675 - 135468199 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0