This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C030037D09RIK SIRNA Assays ›
Gene Symbol:
C030037D09Rik
Gene Name:
RIKEN cDNA C030037D09 gene
Gene Aliases:
9330160M04
Chromosome Location:
Chr.11: 88718643 - 88728572 on Build GRCm38
UniGene ID:
Mm.396842
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n396074

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C030037D09Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038058.1 4 654
GenBank mRNA AK021127.1 - 4 462
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY728261.1 - 4 455 Chr. 11 - 88718842 - 88728363 [+] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0