This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other E230029C05RIK SIRNA Assays ›
Gene Symbol:
E230029C05Rik
Gene Name:
RIKEN cDNA E230029C05 gene
Gene Aliases:
AI452102, B230337J15
Chromosome Location:
Chr.7: 90029159 - 90049071 on Build GRCm38
UniGene ID:
Mm.31753
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n395205

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: E230029C05Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110364.1 6 664
NR_110365.1 6 601
GenBank mRNA AK046045.1 - 6 662
AK157435.1 - 6 600
BE650787.1 - 4 446
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB312740.2 - 5 481 Chr. 7 - 90036993 - 90049069 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0