This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG5 SIRNA Assays ›
Gene Symbol:
Snhg5
Gene Name:
small nucleolar RNA host gene 5
Gene Aliases:
2810026P18Rik, 4933428I09Rik, mU50HG-b
Chromosome Location:
Chr.9: 88521052 - 88522897 on Build GRCm38
UniGene ID:
Mm.21798
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n394360

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snhg5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040721.1 5 667
GenBank mRNA AK012825.1 - 5 660
BC032970.1 - 5 643
KF876690.1 - 5 661
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA231154.1 - 4 200 Chr. 9 - 88521097 - 88522328 [-] on Build GRCm38
GenBankmRNA BY711938.1 - 5 659 Chr. 9 - 88521073 - 88522890 [-] on Build GRCm38
GenBankmRNA EH109332.1 - 4 558 Chr. 9 - 88521372 - 88522587 [-] on Build GRCm38
GenBankmRNA AV445958.1 - 3 168 Chr. 9 - 88521083 - 88521965 [-] on Build GRCm38
GenBankmRNA CJ180500.1 - 2 274 Chr. 9 - 88521243 - 88521869 [-] on Build GRCm38
GenBankmRNA CA539769.1 - 5 369 Chr. 9 - 88521162 - 88522587 [-] on Build GRCm38
GenBankmRNA AA538410.1 - 5 429 Chr. 9 - 88521250 - 88522660 [-] on Build GRCm38
GenBankmRNA CB194977.1 - 6 385 Chr. 9 - 88521064 - 88522879 [-] on Build GRCm38
GenBankmRNA CA979224.1 - 5 632 Chr. 9 - 88521143 - 88522862 [-] on Build GRCm38
GenBankmRNA BG802512.1 - 5 352 Chr. 9 - 88521233 - 88522582 [-] on Build GRCm38
GenBankmRNA BE646905.1 - 3 256 Chr. 9 - 88521148 - 88522053 [-] on Build GRCm38
GenBankmRNA AV467957.1 - 5 284 Chr. 9 - 88521188 - 88522879 [-] on Build GRCm38
GenBankmRNA AV497822.1 - 5 328 Chr. 9 - 88521246 - 88522558 [-] on Build GRCm38
GenBankmRNA CD563600.1 - 5 350 Chr. 9 - 88521139 - 88522579 [-] on Build GRCm38
GenBankmRNA AV472159.1 - 6 380 Chr. 9 - 88521334 - 88522872 [-] on Build GRCm38
GenBankmRNA AV455592.1 - 5 423 Chr. 9 - 88521303 - 88522632 [-] on Build GRCm38
GenBankmRNA BI440435.1 - 6 375 Chr. 9 - 88521221 - 88522866 [-] on Build GRCm38
GenBankmRNA AV463648.1 - 5 374 Chr. 9 - 88521329 - 88522586 [-] on Build GRCm38
GenBankmRNA DV055865.1 - 413 Chr. - - [-] on Build

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0