This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG5 SIRNA Assays ›
Gene Symbol:
Snhg5
Gene Name:
small nucleolar RNA host gene 5
Gene Aliases:
2810026P18Rik, 4933428I09Rik, mU50HG-b
Chromosome Location:
Chr.9: 88521052 - 88522897 on Build GRCm38
UniGene ID:
Mm.21798
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n394359

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snhg5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040721.1 1 382
GenBank mRNA AK012825.1 - 1 375
BC032970.1 - 1 358
KF876690.1 - 1 376
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA EH109332.1 - 1 72 Chr. 9 - 88521372 - 88522587 [-] on Build GRCm38
GenBankmRNA CX564949.1 - 1 363 Chr. 9 - 88521402 - 88522879 [-] on Build GRCm38
GenBankmRNA AV577079.1 - 1 356 Chr. 9 - 88521930 - 88522871 [-] on Build GRCm38
GenBankmRNA AA538410.1 - 1 144 Chr. 9 - 88521250 - 88522660 [-] on Build GRCm38
GenBankmRNA BU937714.1 - 2 153 Chr. 9 - 88521050 - 88522929 [-] on Build GRCm38
GenBankmRNA CA983870.1 - 1 363 Chr. 9 - 88521105 - 88522878 [-] on Build GRCm38
GenBankmRNA CA979224.1 - 1 347 Chr. 9 - 88521143 - 88522862 [-] on Build GRCm38
GenBankmRNA BG802512.1 - 1 67 Chr. 9 - 88521233 - 88522582 [-] on Build GRCm38
GenBankmRNA BG803337.1 - 1 342 Chr. 9 - 88521423 - 88522857 [-] on Build GRCm38
GenBankmRNA BG805289.1 - 1 342 Chr. 9 - 88521423 - 88522857 [-] on Build GRCm38
GenBankmRNA AV497822.1 - 1 43 Chr. 9 - 88521246 - 88522558 [-] on Build GRCm38
GenBankmRNA AV455592.1 - 1 138 Chr. 9 - 88521303 - 88522632 [-] on Build GRCm38
GenBankmRNA BM503554.1 - 1 332 Chr. 9 - 88521650 - 88522847 [-] on Build GRCm38
GenBankmRNA AV463648.1 - 1 89 Chr. 9 - 88521329 - 88522586 [-] on Build GRCm38
GenBankmRNA DV055865.1 - 128 Chr. - - [-] on Build
GenBankmRNA BY711938.1 - 1 374 Chr. 9 - 88521073 - 88522890 [-] on Build GRCm38
GenBankmRNA BX527624.1 - 1 359 Chr. 9 - 88521924 - 88522873 [-] on Build GRCm38
GenBankmRNA AV449185.1 - 1 336 Chr. 9 - 88521936 - 88522851 [-] on Build GRCm38
GenBankmRNA CA322063.1 - 1 347 Chr. 9 - 88521403 - 88522862 [-] on Build GRCm38
GenBankmRNA AV480182.1 - 1 354 Chr. 9 - 88521925 - 88522864 [-] on Build GRCm38
GenBankmRNA CA539769.1 - 1 84 Chr. 9 - 88521162 - 88522587 [-] on Build GRCm38
GenBankmRNA BQ257696.1 - 2 100 Chr. 9 - 88521660 - 88522869 [-] on Build GRCm38
GenBankmRNA CB194977.1 - 2 100 Chr. 9 - 88521064 - 88522879 [-] on Build GRCm38
GenBankmRNA AV452005.1 - 1 265 Chr. 9 - 88521652 - 88522780 [-] on Build GRCm38
GenBankmRNA CD563600.1 - 1 65 Chr. 9 - 88521139 - 88522579 [-] on Build GRCm38
GenBankmRNA AV492898.1 - 1 123 Chr. 9 - 88521302 - 88522640 [-] on Build GRCm38
GenBankmRNA AV472159.1 - 2 95 Chr. 9 - 88521334 - 88522872 [-] on Build GRCm38
GenBankmRNA AV484128.1 - 1 341 Chr. 9 - 88521921 - 88522859 [-] on Build GRCm38
GenBankmRNA BI440435.1 - 2 90 Chr. 9 - 88521221 - 88522866 [-] on Build GRCm38
GenBankmRNA CB056626.1 - 1 349 Chr. 9 - 88521409 - 88522864 [-] on Build GRCm38
GenBankmRNA CA467755.1 - 1 410 Chr. 9 - 88521503 - 88522888 [-] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header