This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG5 SIRNA Assays ›
Gene Symbol:
Snhg5
Gene Name:
small nucleolar RNA host gene 5
Gene Aliases:
2810026P18Rik, 4933428I09Rik, mU50HG-b
Chromosome Location:
Chr.9: 88521052 - 88522897 on Build GRCm38
UniGene ID:
Mm.21798
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n394355

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snhg5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040721.1 1 319
GenBank mRNA AK012825.1 - 1 312
BC032970.1 - 1 295
KF876690.1 - 1 313
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY711938.1 - 1 311 Chr. 9 - 88521073 - 88522890 [-] on Build GRCm38
GenBankmRNA CX564949.1 - 1 300 Chr. 9 - 88521402 - 88522879 [-] on Build GRCm38
GenBankmRNA BX527624.1 - 1 296 Chr. 9 - 88521924 - 88522873 [-] on Build GRCm38
GenBankmRNA CA322063.1 - 1 284 Chr. 9 - 88521403 - 88522862 [-] on Build GRCm38
GenBankmRNA CA983870.1 - 1 300 Chr. 9 - 88521105 - 88522878 [-] on Build GRCm38
GenBankmRNA BG803337.1 - 1 279 Chr. 9 - 88521423 - 88522857 [-] on Build GRCm38
GenBankmRNA CA979224.1 - 1 284 Chr. 9 - 88521143 - 88522862 [-] on Build GRCm38
GenBankmRNA BG805289.1 - 1 279 Chr. 9 - 88521423 - 88522857 [-] on Build GRCm38
GenBankmRNA BM503554.1 - 1 269 Chr. 9 - 88521650 - 88522847 [-] on Build GRCm38
GenBankmRNA CB056626.1 - 1 286 Chr. 9 - 88521409 - 88522864 [-] on Build GRCm38
GenBankmRNA CA467755.1 - 1 347 Chr. 9 - 88521503 - 88522888 [-] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0