This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4833411C07RIK SIRNA Assays ›
Gene Symbol:
4833411C07Rik
Gene Name:
RIKEN cDNA 4833411C07 gene
Gene Aliases:
-
Chromosome Location:
Chr.8: 10899922 - 10902334 on Build GRCm38
UniGene ID:
Mm.139075
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n394203

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4833411C07Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_102285.1 2 355
GenBank mRNA AK014677.1 - 2 266
AK050245.1 - 2 356
BC052524.1 - 349
BY714173.1 - 2 266
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB653859.1 - 2 356 Chr. 8 - 10899922 - 10901846 [+] on Build GRCm38
GenBankmRNA AI099118.1 - 2 337 Chr. 8 - 10899941 - 10901773 [+] on Build GRCm38
GenBankmRNA AI116042.1 - 2 178 Chr. 8 - 10900099 - 10901736 [+] on Build GRCm38
GenBankmRNA BG175492.1 - 2 416 Chr. 8 - 10899894 - 10901873 [+] on Build GRCm38
GenBankmRNA AI286405.1 - 2 354 Chr. 8 - 10899923 - 10900477 [+] on Build GRCm38
GenBankmRNA AI529309.1 - 2 354 Chr. 8 - 10899923 - 10900496 [+] on Build GRCm38
GenBankmRNA BE689236.1 - 2 340 Chr. 8 - 10899937 - 10901664 [+] on Build GRCm38
GenBankmRNA EL956828.1 - 2 345 Chr. 8 - 10899935 - 10901650 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0