This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4833411C07RIK SIRNA Assays ›
Gene Symbol:
4833411C07Rik
Gene Name:
RIKEN cDNA 4833411C07 gene
Gene Aliases:
-
Chromosome Location:
Chr.8: 10899922 - 10902334 on Build GRCm38
UniGene ID:
Mm.139075
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n394201

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4833411C07Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_102285.1 1 211
GenBank mRNA AK014677.1 - 1 122
AK050245.1 - 1 212
BC052524.1 - 205
BY714173.1 - 1 122
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB653859.1 - 1 212 Chr. 8 - 10899922 - 10901846 [+] on Build GRCm38
GenBankmRNA BX523393.1 - 1 173 Chr. 8 - 10899959 - 10901671 [+] on Build GRCm38
GenBankmRNA AI099118.1 - 1 193 Chr. 8 - 10899941 - 10901773 [+] on Build GRCm38
GenBankmRNA AI116042.1 - 1 34 Chr. 8 - 10900099 - 10901736 [+] on Build GRCm38
GenBankmRNA BG175492.1 - 1 272 Chr. 8 - 10899894 - 10901873 [+] on Build GRCm38
GenBankmRNA AI286405.1 - 1 210 Chr. 8 - 10899923 - 10900477 [+] on Build GRCm38
GenBankmRNA AI529309.1 - 1 210 Chr. 8 - 10899923 - 10900496 [+] on Build GRCm38
GenBankmRNA BE689236.1 - 1 196 Chr. 8 - 10899937 - 10901664 [+] on Build GRCm38
GenBankmRNA EL956828.1 - 1 201 Chr. 8 - 10899935 - 10901650 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0