siRNA Details
Gene Symbol: A630023A22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041587.1 | - | 5 | 863 | ||
AW989347.1 | - | 2 | 201 |

Gene Symbol: A630023A22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041587.1 | - | 5 | 863 | ||
AW989347.1 | - | 2 | 201 |