siRNA Details
Gene Symbol: E330018M18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087768.1 | - | 2 | 641 | ||
AK141748.1 | - | 2 | 610 |

Gene Symbol: E330018M18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087768.1 | - | 2 | 641 | ||
AK141748.1 | - | 2 | 610 |