This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 2900040C04RIK SIRNA Assays ›
Gene Symbol:
2900040C04Rik
Gene Name:
RIKEN cDNA 2900040C04 gene
Gene Aliases:
-
Chromosome Location:
Chr.14: 76251265 - 76252334 on Build GRCm38
UniGene ID:
Mm.233565
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n389843

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2900040C04Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK013627.1 - 2 179
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU843427.1 - 2 127 Chr. 14 - 76251509 - 76252327 [+] on Build GRCm38
GenBankmRNA BY712593.1 - 2 176 Chr. 14 - 76251462 - 76252325 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0