This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 6720468P15RIK SIRNA Assays ›
Gene Symbol:
6720468P15Rik
Gene Name:
RIKEN cDNA 6720468P15 gene
Gene Aliases:
-
Chromosome Location:
Chr.19: 57508564 - 57512788 on Build GRCm38
UniGene ID:
Mm.12411
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n387851

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 6720468P15Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040306.1 4 1143
NR_040307.1 3 781
GenBank mRNA AK020151.1 - 4 1121
AK040090.1 - 4 1084
AK078497.1 - 4 1144
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CF582705.1 - 3 791 Chr. 19 - 57509066 - 57512279 [+] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0