siRNA Details
Gene Symbol: 2700089I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045308.1 | 5 | 1571 | |||
GenBank mRNA | AK012591.1 | - | 5 | 1572 |

Gene Symbol: 2700089I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045308.1 | 5 | 1571 | |||
GenBank mRNA | AK012591.1 | - | 5 | 1572 |