siRNA Details
Gene Symbol: Gm6602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045362.1 | 1 | 20 | |||
GenBank mRNA | AK090313.1 | - | 1 | 21 |

Gene Symbol: Gm6602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045362.1 | 1 | 20 | |||
GenBank mRNA | AK090313.1 | - | 1 | 21 |