siRNA Details
Gene Symbol: 4930405D11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045956.1 | 2, 3 | 185 | |||
GenBank mRNA | AK147079.1 | - | 2, 3 | 186 |

Gene Symbol: 4930405D11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045956.1 | 2, 3 | 185 | |||
GenBank mRNA | AK147079.1 | - | 2, 3 | 186 |