siRNA Details
Gene Symbol: 2610020C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038156.1 | 2 | 433 | |||
GenBank mRNA | AK011479.1 | - | 2 | 426 | ||
AK029005.1 | - | 2 | 435 |

Gene Symbol: 2610020C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038156.1 | 2 | 433 | |||
GenBank mRNA | AK011479.1 | - | 2 | 426 | ||
AK029005.1 | - | 2 | 435 |