siRNA Details
Gene Symbol: Gm14168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050354.1 | - | 2 | 1861 | ||
BB667779.1 | - | 1 | 375 |

Gene Symbol: Gm14168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050354.1 | - | 2 | 1861 | ||
BB667779.1 | - | 1 | 375 |