This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM19412 SIRNA Assays ›
Gene Symbol:
Gm19412
Gene Name:
predicted gene, 19412
Gene Aliases:
-
Chromosome Location:
Chr.17: 33919289 - 33920485 on Build GRCm38
UniGene ID:
Mm.392082
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n379954

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm19412
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK140218.1 - 2 525
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CJ060377.1 - 2 525 Chr. 17 - 33919400 - 33920485 [-] on Build GRCm38
GenBankmRNA CN833277.1 - 2 409 Chr. 17 - 33919305 - 33920366 [-] on Build GRCm38
GenBankmRNA CF197840.1 - 2 371 Chr. 17 - 33919318 - 33920331 [-] on Build GRCm38
GenBankmRNA EL957151.1 - 2 387 Chr. 17 - 33919397 - 33920343 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0