siRNA Details
Gene Symbol: 1600025M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038168.1 | 3 | 378 | |||
GenBank mRNA | AA098421.1 | - | 3 | 377 | ||
AK005539.1 | - | 3 | 333 |

Gene Symbol: 1600025M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038168.1 | 3 | 378 | |||
GenBank mRNA | AA098421.1 | - | 3 | 377 | ||
AK005539.1 | - | 3 | 333 |